Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CERILLIANT™ MSTFA Reagent, Analytical Standard, MilliporeSigma™ Supelco™

Catalog No. 111017122 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
10 x 1.2 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-7122 10 x 1.2 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-7122 Supplier MilliporeSigma Supelco Supplier No. M13210X1.2ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Specifications

Packaging Ampule
CAS 24589-78-4
Product Line CERILLIANT
Vapor Pressure 8.8 mmHg (27°C)
Vapor Density >1 (vs. air)
Density 1.075 g/mL (at 25°C)
Physical Form Neat
Linear Formula CF3CON(CH3)Si(CH3)3
MDL Number MFCD00000411
Boiling Point 130°C to 132°C
Synonym N-Methyl-N-(trimethylsilyl)trifluoroacetamide; N-Trimethylsilyl-N-methyl trifluoroacetamide; MSTFA
Quantity 10 x 1.2 mL
Formula Weight 199.25
Grade Analytical Standard
Show More Show Less

CERILLIANT is a registered trademark of Cerilliant Corporation

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.