Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CERILLIANT™ MTBSTFA (with 1 t-BDMCS), Analytical Standard, MilliporeSigma™ Supelco™

Catalog No. 111016627 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
5 x 1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-6627 5 x 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-6627 Supplier MilliporeSigma Supelco Supplier No. M1085X1ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Specifications

Packaging Ampule
CAS 77377-52-7
Product Line CERILLIANT
Physical Form Neat
Linear Formula C9H18F3NOSi
MDL Number MFCD00009671
Synonym N-tert-Butyldimethylsilyl-N-methyltrifluoroacetamide with 1% tert-Butyldimethylchlorosilane; N-Methyl-N-tert-butyldimethylsilyltrifluoroacetamide; MTBSTFA + 1% TBDMSCl; Silylating mixture VI
Quantity 5 x 1 mL
Formula Weight 241.33
Grade Analytical Standard

CERILLIANT is a registered trademark of Cerilliant Corporation

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.