Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CERILLIANT™ Multi-Component Alcohol Mix 100, 100 μg/mL each component in H2O, Sold by MilliporeSigma™ Supelco™

Catalog No. 01258411 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Grade:
Certified Reference Material
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
01-258-411 Certified Reference Material
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-258-411 Supplier MilliporeSigma Supelco Supplier No. A0761.2ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

A multicomponent calibration kit containing acetone, ethanol, isopropanol, and methanol at a concentration of 100 μg/mL. This kit is suitable for calibration of blood alcohol testing methods using GC Headspace.

TRUSTED_SUSTAINABILITY

Specifications

Packaging Ampoule
Product Line CERILLIANT
Physical Form Liquid
Concentration 100 μg/mL each component in H2O
Synonym Multi-component Alcohol Mix
Industry Type Forensics, Toxicology
Grade Certified Reference Material

CERILLIANT™ is a registered trademark of Cerilliant Corporation

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.