Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CERILLIANT™ N-Desethylamiodarone Hydrochloride Solution, 1.0 mg/mL in Methanol (as free base), Sold by MilliporeSigma™ Supelco™

Certified reference material

Supplier:  MilliporeSigma Supelco D0551ML

Catalog No. 01-001-178


Add to Cart

Description

Description

A Certified Spiking Solution™ for use in therapeutic drug monitoring analyses by HPLC or LCMS. N-Desethylamiodarone is the primary, active metabolite of amiodarone, an antiarrhythmic agent marketed as Pacerone™, Cordarone™, Aratac, and Atlansil and monitored by clinical labs to ensure patients remain within the drugs therapeutic range.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

67-56-1 , 96027-74-6
MFCD08060094
1.0 mg/mL in methanol (as free base)
Ampule
9.7°C (49.5°F) - closed cup
C23H26ClI2NO3
UN 1230 - class 3 - PG 2 - Methanol, solution
653.72
1 mL
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

H331-H370

H311

EINECSNumber : 200-659-6

Recommended Storage : -20°C

SDS
Documents

Documents

Product Certifications
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

CERILLIANT is a registered trademark of Cerilliant Corporation