Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CERILLIANT™ N-Desmethyl U-47700 Solution, 1 mg/mL in Acetonitrile, Sold by MilliporeSigma™ Supelco™

Catalog No. 01001243 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-001-243 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-001-243 Supplier MilliporeSigma Supelco Supplier No. D1671ML
 Restricted Item
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Certified reference material

N-Desmethyl U-47700 is the primary urinary metabolite of U-47700. A certified Snap-N-Spike™ solution suitable for use as starting material in calibrators and controls in N-Desmethyl U-47700 LC/MS or GC/MS testing methods for clinical toxicology, urine drug testing or forensic analysis applications.
TRUSTED_SUSTAINABILITY

Specifications

CAS 75-05-8, 67579-73-1
Linear Formula C15H20Cl2N2O
UN Number UN 1648 3 / PGII
Concentration 1 mg/mL in acetonitrile
Formula Weight 315.24
Packaging Ampule
Quantity 1 mL
Flash Point 2°C (35.6°F) - closed cup

CERILLIANT is a registered trademark of Cerilliant Corporation

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.