Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CERILLIANT™ N-Desmethylcitalopram-D3 Hydrochloride Solution, 100 μg/mL in Methanol, Sold by MilliporeSigma™ Supelco™

Certified reference material

Supplier:  MilliporeSigma Supelco D0741ML

Catalog No. 01-001-194


Add to Cart

Description

Description

A stable-labeled internal standard of N-Desmethylcitalopram, an active metabolite of the SSRI antidepressant drug citalopram. Citalopram, marketed as Celexa™ in the US and Canada, is used to treat major depression associated with mood disorders.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

67-56-1
MFCD28138332
100 μg/mL in methanol (as free base)
349.84
1 mL
C19D3H16FN2O · HCl
UN 1230 - class 3 - PG 2 - Methanol, solution
N-Desmethylcitalopram-D3 hydrochloride solution
Ampule
9.7°C (49.5°F) - closed cup
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

H331-H370

H311

EINECSNumber : 200-659-6

Recommended Storage : -20°C

SDS
Documents

Documents

Product Certifications
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

CERILLIANT is a registered trademark of Cerilliant Corporation