Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CERILLIANT™ N-Desmethylclobazam-13C6 Solution, 100 μg/mL in Acetonitrile, Sold by MilliporeSigma™ Supelco™

Catalog No. 01001233 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-001-233 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-001-233 Supplier MilliporeSigma Supelco Supplier No. D1281ML
Add to Cart
Add to Cart

Certified reference material

N-Desmethylclobazam-13C6 is a stable-labeled structural isomer of N-Desmethylclobazam, also known as norclobazam. This standard is suitable for quantification of N-Desmethylclobazam levels in urine or other matrices by LC/MS or GC/MS for clinical toxicology, urine drug testing, or forensic analysis applications. Norclobazam is a pharmacologically active biomarker and a major metabolite of clobazam, a benzodiazepine for use as an anxiolytic and anticonvulsant.
TRUSTED_SUSTAINABILITY

Specifications

CAS 75-05-8
Linear Formula 13C6C9H11ClN2O2
MDL Number MFCD29036792
UN Number UN 1648 3 / PGII
Concentration 100 μg/mL in acetonitrile
Formula Weight 292.67
Packaging Ampule
Quantity 1 mL

CERILLIANT is a registered trademark of Cerilliant Corporation

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.