Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CERILLIANT™ N-Desmethylclozapine-D8 Solution, 100 μg/mL in Methanol, Sold by MilliporeSigma™ Supelco™

Certified reference material

Supplier:  MilliporeSigma Supelco D1691ML

Catalog No. 01-001-245


Add to Cart

Description

Description

N-Desmethylclozapine is an active metabolite of Clozapine, an atypical antipsychotic drug used for treatment of schizophrenia. Clozapine is sold under the trade names Clozaril and FrazaClo in the US. This Snap-N-Shoot™ stable-labeled internal standard is suitable for quantitation of N-desmethylclozapine levels in urine, serum, or plasma by LC/MS or GC/MS in clinical toxicology, forensic analysis, and urine drug testing applications.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

67-56-1 , 1189888-77-4
UN 1230 - class 3 - PG 2 - Methanol, solution
320.85
1 mL
C17H9D8ClN4
100 μg/mL in methanol
Ampule
9.7°C (49.5°F) - closed cup
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

H331-H370

H311

Recommended Storage : -20°C

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

CERILLIANT is a registered trademark of Cerilliant Corporation