Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CERILLIANT™ N-Desmethylcyclobenzaprine Hydrochloride Solution, 1.0 mg/mL in Methanol, Sold by MilliporeSigma™ Supelco™

Catalog No. 01001208 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-001-208 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-001-208 Supplier MilliporeSigma Supelco Supplier No. D0881ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Certified reference material

A certified Snap-N-Spike™ Solution suitable for cyclobenzaprine testing methods by GC/MS or LC/MS for applications in pain prescription monitoring, forensic analysis, urine drug testing, or clinical toxicology. Also known as norcyclobenzaprine, N-desmethylcyclobenzaprine is the major urinary metabolite of the skeletal muscle relaxant cyclobenzaprine. Cyclobenzaprine, sold under trade names Flexeril™, Fexmid™, and Amrix™, is used to relieve skeletal muscle spasms and associated pain from acute musculoskeletal conditions.
TRUSTED_SUSTAINABILITY

Specifications

CAS 67-56-1
Linear Formula C19H19N · HCl
MDL Number MFCD01731061
UN Number UN 1230 - class 3 - PG 2 - Methanol, solution
Concentration 1.0 mg/mL in methanol (as free base)
Formula Weight 297.82
Packaging Ampule
Quantity 1 mL
Flash Point 9.7°C (49.5°F)

CERILLIANT is a registered trademark of Cerilliant Corporation

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.