Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CERILLIANT™ N-Desmethyltapentadol Solution, 1.0 mg/mL in Methanol, Sold by MilliporeSigma™ Supelco™

Catalog No. 01001176 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-001-176 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-001-176 Supplier MilliporeSigma Supelco Supplier No. D0521ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Certified reference material

N-Desmethyltapentadol is a primary urinary and blood metabolite of the analgesic tapentadol (Nucynta™). This certified reference solution is suitable for numerous GC/MS or LC/MS applications from pharmaceutical testing and prescription pain monitoring to forensic analysis or clinical toxicology. Tapentadol is prescribed for treatment of both acute and chronic pain.
TRUSTED_SUSTAINABILITY

Specifications

CAS 67-56-1, 1246819-18-0
Linear Formula C13H21NO
MDL Number MFCD16294728
UN Number UN 1230 - class 3 - PG 2 - Methanol, solution
Concentration 1.0 mg/mL in methanol
Formula Weight 207.31
Packaging Ampule
Quantity 1 mL
Flash Point 9.7°C (49.5°F) - closed cup

CERILLIANT is a registered trademark of Cerilliant Corporation

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.