Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CERILLIANT™ N-Ethylpentylone-D5 Hydrochloride Solution, 100 μg/mL in Methanol (as free base), Sold by MilliporeSigma™ Supelco™

Certified reference material

Supplier:  MilliporeSigma Supelco E1301ML

Catalog No. 01-001-322


Add to Cart

Description

Description

N-Ethylpentylone, also known as Ephylone, is a stimulant and designer drug found in 'bath salts'. These synthetic cathinones offer recreational uses that mimic the effects of illegal drugs such as cocaine, methamphetamine, and LSD. An internal standard suitable for quantification of N-Ethylpentylone in synthetic cathinone testing methods by LC/MS or GC/MS applications in forensic analysis, urine drug testing, or clinical toxicology.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

67-56-1
UN 1230 - class 3 - PG 2 - Methanol, solution
290.80
1 mL
C14H14D5NO3 · HCl
100 μg/mL in methanol (as free base)
Ampule
9.7°C (49.5°F) - closed cup
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

H331-H370

H311

Recommended Storage : -20°C

SDS
Documents

Documents

Product Certifications
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

CERILLIANT is a registered trademark of Cerilliant Corporation