Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CERILLIANT™ N,N-Dimethyltryptamine (DMT) Solution, 1.0 mg/mL in Methanol, Sold by MilliporeSigma™ Supelco™

Certified reference material

Supplier:  MilliporeSigma Supelco D1021ML

Catalog No. 01-001-216


Add to Cart

Description

Description

This certified solution standard is suitable for N,N-Dimethyltryptamine (DMT) testing methods by GC/MS or LC/MS in clinical toxicology, urine drug testing, or forensic analysis applications. DMT is a naturally occurring hallucinogenic tryptamine derived from the amino acid tryptophan. Abuse of DMT reportedly can cause auditory and visual hallucinations, and mild stimulant effects such as increased heart rate and blood pressure.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

67-56-1 , 61-50-7
MFCD00055989
1.0 mg/mL in methanol
Ampule
9.7°C (49.5°F) - closed cup
C12H16N2
UN 1230 - class 3 - PG 2 - Methanol, solution
188.27
1 mL
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

H331-H370

H311

Recommended Storage : -20°C

SDS
Documents

Documents

Product Certifications
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

CERILLIANT is a registered trademark of Cerilliant Corporation