Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CERILLIANT™ Naloxone-D5-3-beta-D-glucuronide Solution, 100 μg/mL in Methanol:Water (9:1), Sold by MilliporeSigma™ Supelco™

Catalog No. 01001765 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-001-765 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-001-765 Supplier MilliporeSigma Supelco Supplier No. N1091ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Certified reference material

Naloxone-3-β-D-glucuronide is a major urinary metabolite of naloxone, an opiate drug used to counter the effects of opioid overdose. This stable-labeled internal standard of naloxone-3-β-D-glucuronide is suitable for quantitation of naloxone glucuronide metabolite levels using LC-MS/MS or GC/MS for synthetic opiate testing methods in forensic analysis, clinical toxicology, or urine drug testing.
TRUSTED_SUSTAINABILITY

Specifications

CAS 67-56-1
Linear Formula C25H24D5NO10
UN Number UN 1230 - class 3 - PG 2 - Methanol, solution
Concentration 100 μg/mL in methanol:water (9:1)
Formula Weight 508.53
Packaging Ampule
Quantity 1 mL
Flash Point 9.7°C (49.5°F) - closed cup

CERILLIANT is a registered trademark of Cerilliant Corporation

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.