Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CERILLIANT™ Naloxone-D5 Solution, 100 μg/mL in Methanol, Sold by MilliporeSigma™ Supelco™

Catalog No. 01001734 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-001-734 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-001-734 Supplier MilliporeSigma Supelco Supplier No. N0631ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Certified reference material

A new internal standard suitable for numerous LC/MS and GC/MS applications from forensic analysis and clinical toxicology testing to pharmaceutical analysis and isotope dilution methods. Naloxone is an opiate drug used to counter the effects of opioid overdose.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Naloxol
CAS 67-56-1, 1261079-38-2
Concentration 100 μg/mL in methanol
Packaging Ampule
Linear Formula C19H16NO4D5
MDL Number MFCD08063611
Quantity 1 mL
UN Number UN 1230 - class 3 - PG 2 - Methanol, solution
Flash Point 9.7°C (49.5°F) - closed cup
Molecular Weight (g/mol) 329.39
Formula Weight 332.41
Show More Show Less

CERILLIANT is a registered trademark of Cerilliant Corporation

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.