Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CERILLIANT™ Naloxone N-oxide Solution, 100 μg/mL in Acetonitrile:Water (1:1), Sold by MilliporeSigma™ Supelco™

Catalog No. 01001520 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-001-520 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-001-520 Supplier MilliporeSigma Supelco Supplier No. IMPN007041ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Certified reference material

Naloxone N-oxide is an impurity in pharmaceutical drug formulations of naloxone as well as a potential naloxone urinary metabolite. This certified solution standard is suitable for numerous LC or LC/MS applications from pharmaceutical testing and research to clinical toxicology or forensic analysis. Naloxone, sold under various trade names including Narcan™, Nalone, and Narcanti, is an opiate drug used to counter the effects of opiate overdose such as those involving heroin or morphine.
TRUSTED_SUSTAINABILITY

Specifications

CAS 75-05-8, 112242-14-5
Linear Formula C19H21NO5
UN Number UN 1648 3 / PGII
Concentration 100 μg/mL in acetonitrile:water (1:1)
Synonym Naloxone N-oxide solution
Formula Weight 343.37
Packaging Ampule
Quantity 1 mL
Flash Point 13°C (55.4°F) - closed cup

CERILLIANT is a registered trademark of Cerilliant Corporation

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.