Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CERILLIANT™ Naltrexone-3-beta-D-glucuronide Solution, 100 μg/mL in Methanol, Sold by MilliporeSigma™ Supelco™

Certified reference material

Supplier:  MilliporeSigma Supelco N1061ML

Catalog No. 01-001-763


Add to Cart

Description

Description

A certified solution standard of Naltrexone-3-β-D-glucuronide, a metabolite of the opiate Naltrexeone. Sold under the trade names Revia, Depade and Vivitrol, naltrexone is prescribed primarily for the management of alcohol or opiate dependence. This analytical reference standard is applicable for use in LC/MS or GC/MS applications such as pharmaceutical research, forensic analysis, clinical toxicology, pain prescription monitoring or urine drug testing.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

67-56-1 , 76630-71-2
UN 1230 - class 3 - PG 2 - Methanol, solution
517.53
1 mL
C26H31NO10
100 μg/mL in methanol
Ampule
9.7°C (49.5°F) - closed cup
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

H331-H370

H311

Recommended Storage : -20°C

SDS
Documents

Documents

Product Certifications
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

CERILLIANT is a registered trademark of Cerilliant Corporation