Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CERILLIANT™ Naltrexone-D3 Solution, 100 μg/mL in Methanol, Sold by MilliporeSigma™ Supelco™

Catalog No. 01001721 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-001-721 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-001-721 Supplier MilliporeSigma Supelco Supplier No. N0471ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Certified reference material

Naltrexone is an opiate sold under the trade names Revia, Depade and Vivitrol™ and is prescribed primarily for the management of alcohol or opiate dependence. This internal standard is suitable for Naltrexol testing or isotope dilution methods in pharmaceutical research, forensic analysis, clinical toxicology, pain prescription monitoring or urine drug testing.
TRUSTED_SUSTAINABILITY

Specifications

CAS 67-56-1, 1261080-26-5
Linear Formula C20H20NO4D3
UN Number UN 1230 - class 3 - PG 2 - Methanol, solution
Concentration 100 μg/mL in methanol
Formula Weight 344.42
Packaging Ampule
Quantity 1 mL
Flash Point 9.7°C (49.5°F) - closed cup

CERILLIANT is a registered trademark of Cerilliant Corporation

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.