Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CERILLIANT™ Naphyrone-D5 Hydrochloride Solution, 100 μg/mL in Methanol (as free base), Sold by MilliporeSigma™ Supelco™

Catalog No. 01001750 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-001-750 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-001-750 Supplier MilliporeSigma Supelco Supplier No. N0851ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Certified reference material

Naphyrone is a synthetic cathinone found in numerous bath salt blends under street names such as NRG-1 and Energy-1. The designer drug emerged in late 2010 as a new legal high in the UK after the banning of mephedrone. This Certified Spiking Solution™ is suitable for use as an internal standard in naphyrone testing methods for clinical toxicology, forensic analysis, and urine drug testing applications.
TRUSTED_SUSTAINABILITY

Specifications

CAS 67-56-1
Linear Formula C19H18D5NO · HCl
UN Number UN 1230 - class 3 - PG 2 - Methanol, solution
Concentration 100 μg/mL in methanol (as free base)
Formula Weight 322.88
Packaging Ampule
Quantity 1 mL
Flash Point 9.7°C (49.5°F)

CERILLIANT is a registered trademark of Cerilliant Corporation

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.