Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CERILLIANT™ Naphyrone Hydrochloride, 1.0 mg/mL in Methanol (as free base), Sold by MilliporeSigma™ Supelco™

Catalog No. 01001735 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-001-735 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-001-735 Supplier MilliporeSigma Supelco Supplier No. N0671ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Certified reference material

Also known as NRG-1, Energy-1, or O-2482, naphyrone is a new designer drug and stimulant with cases of abuse reported in Europe. The drug emerged in late 2010 as a new legal high in the UK after the banning of mephedrone. This certified solution standard offers numerous applications in GC/MS or LC/MS testing from forensic analysis and clinical toxicology to urine drug monitoring.
TRUSTED_SUSTAINABILITY

Specifications

CAS 67-56-1, 850352-11-3
Linear Formula C19H23NO · HCl
MDL Number MFCD18379564
UN Number UN 1230 - class 3 - PG 2 - Methanol, solution
Concentration 1.0 mg/mL in methanol (as free base)
Synonym Naphyrone hydrochloride solution, Energy-1, NRG-1, O-2482
Formula Weight 317.85
Packaging Ampule
Quantity 1 mL
Flash Point 9.7°C (49.5°F) - closed cup

CERILLIANT is a registered trademark of Cerilliant Corporation

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.