Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CERILLIANT™ Nicotinamide (Vitamin B3) Solution, 1.0 mg/mL in Methanol, Sold by MilliporeSigma™ Supelco™

Catalog No. 01002143 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-002-143 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-002-143 Supplier MilliporeSigma Supelco Supplier No. V0161ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Certified reference material

Nicotinamide is also known as niacinamide or nicotinic acid amide. It is the water-soluble form of Vitamin B3 found in plant and animal foods and mostly used as a dietary supplement.
TRUSTED_SUSTAINABILITY

Specifications

CAS 67-56-1, 98-92-0
Melting Point 128 to 131°C (lit.)
Linear Formula C6H6N2O
MDL Number MFCD00006395
UN Number UN 1230 - class 3 - PG 2 - Methanol, solution
Concentration 1.0 mg/mL in methanol
Synonym Nicotinamide, Vitamin B3, Niacinamide, Nicotinic acid amide, Pyridine-3-carboxylic acid amide, Vitamin B3, Vitamin PP
Formula Weight 122.12
Packaging Ampule
Quantity 1 mL
Flash Point 9.7°C (49.5°F) - closed cup
Show More Show Less

CERILLIANT is a registered trademark of Cerilliant Corporation

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.