Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CERILLIANT™ Nicotinic acid-13C6 Solution, 100 μg/mL in Methanol, Sold by MilliporeSigma™ Supelco™

Catalog No. 01002158 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-002-158 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-002-158 Supplier MilliporeSigma Supelco Supplier No. V0351ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Certified reference material

Nicotinic acid, also known as niacin, is a water-soluble vitamin of the B complex and an essential human nutrient. Niacin defficiency, often the result of impaired nutritional status associated with conditions ranging from malnutrition to chronic alcoholism, is frequently monitored in patient plasma samples by LC-MS/MS.
TRUSTED_SUSTAINABILITY

Specifications

CAS 67-56-1, 1189954-79-7
Linear Formula 13C6H5NO2
UN Number UN 1230 - class 3 - PG 2 - Methanol, solution
Concentration 100 μg/mL in methanol
Synonym Vitamin B3-13C6 solution
Formula Weight 129.07
Packaging Ampule
Quantity 1 mL
Flash Point 9.7°C (49.5°F) - closed cup

CERILLIANT is a registered trademark of Cerilliant Corporation

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.