Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CERILLIANT™ Nitrazepam Solution, 1.0 mg/mL in Acetonitrile, Sold by MilliporeSigma™ Supelco™

Catalog No. 01001789 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-001-789 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-001-789 Supplier MilliporeSigma Supelco Supplier No. N9061ML
 Restricted Item
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Certified reference material

A certified solution standard suitable for use as starting material in calibrators and controls of nitrazepam for LC/MS or GC/MS testing methods. Nitrazepam is a benzodiazepine sold as a treatment for severe insomnia. This drug is marketed under the trade names Mogadon, Nitrados, and Somnite.
TRUSTED_SUSTAINABILITY

Specifications

CAS 75-05-8, 146-22-5
Linear Formula C15H11N3O3
UN Number UN 1648 3 / PGII
Concentration 1.0 mg/mL in acetonitrile
Formula Weight 281.27
Packaging Ampule
Quantity 1 mL

CERILLIANT is a registered trademark of Cerilliant Corporation

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.