Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CERILLIANT™ Norchlordiazepoxide Solution, 1.0 mg/mL (Acetonitrile:DMSO (80:20)(v/v)), Sold by MilliporeSigma™ Supelco™

Catalog No. 01001759 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-001-759 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-001-759 Supplier MilliporeSigma Supelco Supplier No. N1001ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Certified reference material

Norchlordiazepoxide is a major urinary metabolite of the benzodiazepine chlordiazepoxide, indicated for short term treatment of severe anxiety and for the management of acute alcohol withdrawal syndrome. The metabolite, also known as N-desmethylchlordiazepoxide, is pharmacologically active. This Certified Spiking Solution™ is suitable for GC/MS or LC/MS methods in chlordiazepoxide testing for forensic analysis, urine drug testing, or clinical toxicology.
TRUSTED_SUSTAINABILITY

Specifications

CAS 75-05-8, 7722-15-8
Linear Formula C15H12ClN3O
MDL Number MFCD27976665
UN Number UN 1648 3 / PGII
Concentration 1.0 mg/mL (acetonitrile:DMSO (80:20)(v/v))
Formula Weight 285.73
Packaging Ampule
Quantity 1 mL
Flash Point 2°C (35.6°F) - closed cup

CERILLIANT is a registered trademark of Cerilliant Corporation

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.