Learn More
CERILLIANT™ Norcodeine-d3, 1.0 mg/mL in Methanol, Sold by MilliporeSigma™ Supelco™
Shop All MilliporeSigma Supelco ProductsDescription
Specifications
Specifications
| CAS | 67-56-1 |
| Linear Formula | C17D3H16NO3 |
| UN Number | UN 1230 - class 3 - PG 2 - Methanol, solution |
| Concentration | 1.0 mg/mL in methanol |
| Synonym | Norcodeine-D3 solution |
| Formula Weight | 288.36 |
| Packaging | Ampule |
| Quantity | 1 mL |
| Flash Point | 9.7°C (49.5°F) - closed cup |
CERILLIANT is a registered trademark of Cerilliant Corporation
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.