Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CERILLIANT™ Norfentanyl oxalate Solution, 1.0 mg/mL in Methanol (as free base), Sold by MilliporeSigma™ Supelco™

Catalog No. 01001710 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-001-710 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-001-710 Supplier MilliporeSigma Supelco Supplier No. N0311ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Certified reference material

Norfentanyl is a major urinary metabolite of fentanyl, a potent synthetic analgesic sold under various trade names including Sublimaze, Duragesic™, Actiq™, and Fentora™. This Certified Spiking Solution™ is suitable for numerous GC/MS or LC/MS testing applications from clinical toxicology and forensic analysis to urine drug testing and pain prescription monitoring.
TRUSTED_SUSTAINABILITY

Specifications

Packaging Ampule
CAS 67-56-1
Flash Point 9.7°C (49.5°F) - closed cup
Concentration 1.0 mg/mL in methanol (as free base)
Linear Formula C14H20N2O · C2H2O4
MDL Number MFCD08272799
UN Number UN 1230 - class 3 - PG 2 - Methanol, solution
Quantity 1 mL
Formula Weight 322.36

CERILLIANT is a registered trademark of Cerilliant Corporation

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.