Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CERILLIANT™ Norfluoxetine-D6 oxalate Solution, 100 μg/mL in Methanol (as free base), Sold by MilliporeSigma™ Supelco™

Catalog No. 01001796 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-001-796 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-001-796 Supplier MilliporeSigma Supelco Supplier No. N9221ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Certified reference material

Norfluoxetine is a primary blood metabolite of the selective serotonin reuptake inhibitor (SSRI) fluoxetine, an antidepressant, sold under the trade name Prozac™. This internal standard is suitable for quantitation of fluoxetine levels in urine, serum, or plasma by LC/MS or GC/MS for urine drug testing, clinical toxicology, or forensic analysis.
TRUSTED_SUSTAINABILITY

Specifications

CAS 67-56-1
Linear Formula C16D6H10F3NO · C2H2O4
MDL Number MFCD08276450
UN Number UN 1230 - class 3 - PG 2 - Methanol, solution
Concentration 100 μg/mL in methanol (as free base)
Formula Weight 391.37
Packaging Ampule
Quantity 1 mL
Flash Point 9.7°C (49.5°F) - closed cup

CERILLIANT is a registered trademark of Cerilliant Corporation

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.