Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CERILLIANT™ Norhydromorphone Hydrochloride Solution, 1.0 mg/mL in Methanol (as free base), Sold by MilliporeSigma™ Supelco™

Catalog No. 01001764 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-001-764 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-001-764 Supplier MilliporeSigma Supelco Supplier No. N1071ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Certified reference material

A long-term urinary metabolite of hydromorphone, a very potent opioid analgesic marketed under brand names including Palladone and Dilaudid. Hydromorphone is often used medicinally as an alternative to morphine for treatment of pain or as a cough suppressant for painful coughing resulting from continued bronchial irritation. This certified reference standard is suitable for use as a starting material in calibrators or controls for a variety of LC/MS or GC/MS applications from forensic analysis, clinical toxicology, and pain prescription monitoring to urine drug testing and pharmaceutical research.
TRUSTED_SUSTAINABILITY

Specifications

CAS 67-56-1, 14140-35-3
Linear Formula C16H18ClNO3
UN Number UN 1230 - class 3 - PG 2 - Methanol, solution
Concentration 1.0 mg/mL in methanol (as free base)
Formula Weight 307.77
Packaging Ampule
Quantity 1 mL
Flash Point 9.7°C (49.5°F) - closed cup

CERILLIANT is a registered trademark of Cerilliant Corporation

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.