Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CERILLIANT™ (+/-)-Norketamine-D4 Hydrochloride Solution, 100 μg/mL in Methanol, Sold by MilliporeSigma™ Supelco™

Certified reference material

Supplier:  MilliporeSigma Supelco N0371ML

Catalog No. 01-001-715


Add to Cart

Description

Description

Norketamine is the most prevalent urinary metabolite of ketamine, a club drug and widely used anesthetic sold under trade names including Ketanest™, Ketaset™ and Ketalar™. This stable-labeled internal standard is suitable for quantitation of norketamine levels in GC/MS or LC/MS applications in clinical toxicology, urine drug testing, forensic analysis, clinical or diagnostic testing and isotope dilution methods.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

67-56-1
UN 1230 - class 3 - PG 2 - Methanol, solution
264.18
1 mL
C12D4H11Cl2NO
100 μg/mL in methanol (as free base)
Ampule
9.7°C (49.5°F) - closed cup
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

H331-H370

H311

Recommended Storage : -20°C

SDS
Documents

Documents

Product Certifications
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

CERILLIANT is a registered trademark of Cerilliant Corporation