Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CERILLIANT™ (+/-)-Norketamine Hydrochloride Solution, 1.0 mg/mL in Methanol (as free base), Sold by MilliporeSigma™ Supelco™

Catalog No. 01001714 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-001-714 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-001-714 Supplier MilliporeSigma Supelco Supplier No. N0361ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Certified reference material

Norketamine is the major urinary metabolite of ketamine. This certified Snap-N-Spike™ Solution is suitable for numerous GC/MS or LC/MS applications in forensic analysis, urine drug testing, clinical toxicology, and clinical or diagnostic testing. Ketamine, sold as Ketanest™, Ketaset™ and Ketalar™, is an NMDA receptor antagonist used for the induction and maintenance of general anesthesia in human and veterinary medicine. The drug is often abused at clubs, raves, or parties for its noted hallucinogenic and dissociative properties and prescribed by some doctors as an off-label treatment for depression.
TRUSTED_SUSTAINABILITY

Specifications

CAS 67-56-1, 79499-59-5
Linear Formula C12H15Cl2NO
UN Number UN 1230 - class 3 - PG 2 - Methanol, solution
Concentration 1.0 mg/mL in methanol (as free base)
Formula Weight 260.16
Packaging Ampule
Quantity 1 mL
Flash Point 9.7°C (49.5°F) - closed cup

CERILLIANT is a registered trademark of Cerilliant Corporation

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.