Learn More
CERILLIANT™ Norlidocaine Solution, 1 mg/mL in Methanol, Sold by MilliporeSigma™ Supelco™
Certified reference material
Supplier:  MilliporeSigma Supelco N1241ML
Catalog No. 01-001-776
Add to Cart
Description
Description
Norlidocaine is a primary urinary metabolite of lidocaine, a local anesthetic and antiarrhythmic drug. A Certified Snap-N-Spike™ Solution for use in clinical toxicology, forensic analysis, urine drug testing, or pharmaceutical research applications by LC/MS or GC/MS.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
67-56-1 , 7728-40-7
1 mg/mL in methanol
Ampule
9.7°C (49.5°F) - closed cup
C12H18N2O
206.28
1 mL
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
H331-H370
H311
Recommended Storage : -20°C
SDS
Documents
Documents
Product Certifications
Product Title
Select an issue
CERILLIANT is a registered trademark of Cerilliant Corporation