Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CERILLIANT™ Noroxymorphone Hydrochloride Solution, 1.0 mg/mL in Methanol:DMSO (80:20), Sold by MilliporeSigma™ Supelco™

Certified reference material

Supplier:  MilliporeSigma Supelco N0121ML

Catalog No. 01-001-706


Add to Cart

Description

Description

A certified Snap-N-Spike™ solution of noroxymorphone suitable for use in a variety of GC/MS or LC/MS applications from pain prescription monitoring, clinical toxicology, and pharmaceutical research to forensic analysis and urine drug testing. Noroxymorphone is a major metabolite of oxymorphone in urine. Oxymorphone, sold under trade names such as Opana™, Numorphan™, and Numorphone, is an opiate analgesic indicated for the relief of moderate to severe pain.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

67-56-1 , 33522-95-1
MFCD08272693
1.0 mg/mL in methanol:DMSO (80:20) (as free base)
Ampule
22°C (71.6°F) - closed cup
C16H17NO4·HCl
UN 1230 - class 3 - PG 2 - Methanol, solution
323.77
1 mL
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

H331-H370

H311

Recommended Storage : 2 to 8°C

SDS
Documents

Documents

Product Certifications
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

CERILLIANT is a registered trademark of Cerilliant Corporation