Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CERILLIANT™ Norquetiapine Hydrochloride, 1.0 mg/mL in Methanol (as free base), Sold by MilliporeSigma™ Supelco™

Catalog No. 01001738 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-001-738 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-001-738 Supplier MilliporeSigma Supelco Supplier No. N0701ML
Add to Cart
Add to Cart

Certified reference material

Norquetiapine is a primary active metabolite of quetiapine, marketed as the atypical antipsychotic Seroquel™ for the treatment of depression, bipolar disorder, and schizophrenia. It maintains most of the pharmacological effects of quetiapine with potencies similar to the parent drug.
TRUSTED_SUSTAINABILITY

Specifications

CAS 67-56-1, 753475-15-9
Linear Formula C17H17N3S · HCl
MDL Number MFCD09835377
UN Number UN 1230 - class 3 - PG 2 - Methanol, solution
Concentration 1.0 mg/mL in methanol (as free base)
Synonym Norquetiapine hydrochloride solution
Formula Weight 331.86
Packaging Ampule
Quantity 1 mL
Flash Point 9.7°C (49.5°F) - closed cup

CERILLIANT is a registered trademark of Cerilliant Corporation

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.