Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CERILLIANT™ Norsertraline-13C6 Hydrochloride Solution, 100 μg/mL in Methanol (as free base), Sold by MilliporeSigma™ Supelco™

Catalog No. 01001749 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-001-749 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-001-749 Supplier MilliporeSigma Supelco Supplier No. N0841ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Certified reference material

Norsertraline is a major urinary metabolite and N-desmethyl analog of the SSRI antidepressant sertraline. Marketed under trade names such as Zoloft™ and Lustral, sertraline is used to treat major depressive disorder in adults as well as obsessive-compulsive, panic, and social anxiety disorders in both adults and children. Suitable uses for this certified solution standard include as an internal standard for quantitation of norsertraline levels in urine or isotope dilution methods by LC/MS or GC/MS for clinical toxicology, urine drug testing, or forensic analysis.
TRUSTED_SUSTAINABILITY

Specifications

CAS 67-56-1
Linear Formula 13C6C10H15NCl2 · HCl
MDL Number MFCD08063522
UN Number UN 1230 - class 3 - PG 2 - Methanol, solution
Concentration 100 μg/mL in methanol (as free base)
Formula Weight 334.62
Packaging Ampule
Quantity 1 mL
Flash Point 9.7°C (49.5°F)

CERILLIANT is a registered trademark of Cerilliant Corporation

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.