Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CERILLIANT™ Nortilidine-D3 Hydrochloride Solution, 100 μg/mL in Methanol (as free base), Sold by MilliporeSigma™ Supelco™

Certified reference material

Supplier:  MilliporeSigma Supelco N0621ML

Catalog No. 01-001-733


Add to Cart

Description

Description

A new internal standard suitable for numerous GC/MS and LC/MS applications from clinical toxicology, pain prescription monitoring, and forensic analysis to urine drug testing and isotope dilution methods. Nortilidine is the primary, active metabolite of tilidine, a synthetic opioid analgesic used mainly in Germany, Switzerland and Belgium for treatment of moderate to severe pain, both acute and chronic.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

67-56-1 , 1217648-75-3
UN 1230 - class 3 - PG 2 - Methanol, solution
298.82
1 mL
C16D3H18NO2 · HCl
100 μg/mL in methanol (as free base)
Ampule
9.7°C (49.5°F) - closed cup
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

H331-H370

H311

EINECSNumber : 200-659-6

Recommended Storage : -20°C

SDS
Documents

Documents

Product Certifications
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

CERILLIANT is a registered trademark of Cerilliant Corporation