Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CERILLIANT™ O-Desmethyl-cis-tramadol-D6 Hydrochloride Solution, 100 μg/mL in Methanol, Sold by MilliporeSigma™ Supelco™

Catalog No. 01001180 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-001-180 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-001-180 Supplier MilliporeSigma Supelco Supplier No. D0581ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Certified reference material

O-Desmethyl-cis-tramadol is the primary, active metabolite of tramadol, marketed as the opioid analgesic Ultram™ or Tramal, for the treatment of moderate to moderately severe pain. This new stable-labeled internal standard is suitable for the quantitation of O-Desmethyl-cis-tramadol in biological samples using GCMS or LCMS with numerous applications including forensic or clinical toxicology analysis, urine drug testing, or isotope dilution methods.
TRUSTED_SUSTAINABILITY

Specifications

CAS 67-56-1
Linear Formula C15D6H17NO2 · HCl
MDL Number MFCD08063534
UN Number UN 1230 - class 3 - PG 2 - Methanol, solution
Concentration 100 μg/mL in methanol (as free base)
Formula Weight 291.85
Packaging Ampule
Quantity 1 mL
Flash Point 9.7°C (49.5°F) - closed cup

CERILLIANT is a registered trademark of Cerilliant Corporation

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.