Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CERILLIANT™ Olanzapine-d8, 100 μg/mL in Acetonitrile, Sold by MilliporeSigma™ Supelco™

Certified reference material

Supplier:  MilliporeSigma Supelco O0351ML

Catalog No. 01-001-815


Add to Cart

Description

Description

An internal standard for use in LC/MS or GC/MS applications including clinical toxicology analysis, therapeutic drug monitoring, or isotope dilution methods. Marketed under the trade name Zyprexa™, olanzapine is an atypical antipsychotic drug used in the treatment of schizophrenia and bipolar disorder.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

75-05-8
UN 1648 3 / PGII
Olanzapine-D8 solution
Ampule
C17D8H12N4S
100 μg/mL in acetonitrile
320.48
1 mL
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : 200-835-2

Recommended Storage : -20°C

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

CERILLIANT is a registered trademark of Cerilliant Corporation