Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CERILLIANT™ Oxycodone Solution, 1.0 mg/mL in Methanol, Sold by MilliporeSigma™ Supelco™

Certified reference material

Supplier:  MilliporeSigma Supelco O0021ML

Catalog No. 01-001-799


Add to Cart

Description

Description

This Certified Spiking Solution™ is suitable for numerous applications in GC/MS or LC/MS testing of oxycodone from clinical toxicology and urine drug testing to forensic analysis. Oxycodone is a semi-synthetic opioid analgesic sold under many trade names including OxyContin™ and Percocet™ for treatment of moderate to severe pain. Oxycodone is also sold illicitly under street names such as 'Roxies,' 'Blues,' and 'Perks'.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

67-56-1 , 76-42-6
MFCD00190613
1.0 mg/mL in methanol
Ampule
9.7°C (49.5°F) - closed cup
C18H21NO4
UN 1230 - class 3 - PG 2 - Methanol, solution
315.36
1 mL
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

H331-H370

H311

EINECSNumber : 200-659-6

Recommended Storage : 2 to 8°C

SDS
Documents

Documents

Product Certifications
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

CERILLIANT is a registered trademark of Cerilliant Corporation