Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CERILLIANT™ Pain Management Multi-component Opiate Mixture-13 Solution, 100 μg/mL each, Sold by MilliporeSigma™ Supelco™

Catalog No. 01258400 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-258-400 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-258-400 Supplier MilliporeSigma Supelco Supplier No. P0711ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

A multi-component mix containing opiates methadone, buprenorphine, tramadol, codeine, fentanyl, hydrocodone, meperidine, morphine, naloxone, naltrexone, oxycodone, and oxymorphone. This certified solution mix is suitable for numerous applications from pain prescription monitoring, clinical toxicology, and forensic analysis to urine drug testing.

TRUSTED_SUSTAINABILITY

Specifications

Packaging Ampoule
Product Line CERILLIANT
Physical Form Liquid
Concentration 100 μg/mL each component (10 μg/mL Fentanyl)
Industry Type Forensics, Toxicology
Quantity 1 mL
Grade Certified Reference Material

CERILLIANT™ is a registered trademark of Cerilliant Corporation

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.