Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CERILLIANT™ Paroxetine-D6 maleate Solution, 100 μg/mL in Methanol (as free base), Sold by MilliporeSigma™ Supelco™

Certified reference material

Supplier:  MilliporeSigma Supelco P9151ML

Catalog No. 01-001-911


Add to Cart

Description

Description

Paroxetine, marketed under trade names Paxil™ and Aropax, is an SSRI antidepressant used to treat many conditions in adults from major depression and obsessive-compulsive disorder to several anxiety disorders. This stable-labeled internal standard is suitable for quantification of paroxetine using LC/MS or LC-MS/MS methods for applications in clinical or diagnostic testing, clinical toxicology, or pharmaceutical research.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

67-56-1 , 1435728-64-5
MFCD08276451
100 μg/mL in methanol (as free base)
451.47
1 mL
C19D6H14FNO3 · C4H4O4
UN 1230 - class 3 - PG 2 - Methanol, solution
Paroxetine-D6 maleate solution
Ampule
9.7°C (49.5°F) - closed cup
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

H331-H370

H311

EINECSNumber : 200-659-6

Recommended Storage : -20°C

SDS
Documents

Documents

Product Certifications
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

CERILLIANT is a registered trademark of Cerilliant Corporation