Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CERILLIANT™ (+/-)-Pentazocine-13C3 Hydrochloride, 100 μg/mL in Methanol (as free base), Sold by MilliporeSigma™ Supelco™

Catalog No. 01001858 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-001-858 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-001-858 Supplier MilliporeSigma Supelco Supplier No. P0841ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Certified reference material

Pentazocine is a synthetic opioid analgesic sold under trade names Fortral, Talwin, or Talacen and used to treat moderate to moderately severe pain. Suitable use of this certified Snap-N-Spike™ solution and internal standard includes quantitation of pentazocine levels and isotope dilution methods by GC/MS or LC/MS in applications for clinical toxicology, urine drug testing, forensic analysis or pain prescription monitoring.
TRUSTED_SUSTAINABILITY

Specifications

CAS 67-56-1
Linear Formula 13C3C16H27NO · HCl
UN Number UN 1230 - class 3 - PG 2 - Methanol, solution
Concentration 100 μg/mL in methanol (as free base)
Synonym (+/-)-Pentazocine-13C3 hydrochloride solution
Formula Weight 324.86
Packaging Ampule
Quantity 1 mL
Flash Point 9.7°C (49.5°F) - closed cup

CERILLIANT is a registered trademark of Cerilliant Corporation

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.