Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CERILLIANT™ PEth 16:0/18:1-D5 Solution, 100 μg/mL in Methanol (as free phosphate), Sold by MilliporeSigma™ Supelco™

Catalog No. 01001902 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-001-902 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-001-902 Supplier MilliporeSigma Supelco Supplier No. P1511ML

missing translation for 'validMainframeMsg'

Certified reference material

This stable-labeled internal standard is suitable for quantitation of PEth 16:0/18:1 in LC/MS or GC/MS applications. PEth 16:0/18:1, also known as phosphatidylethanol 16:0/18:1, is a sensitive and specific biomarker for alcohol use and abuse. Since the detection window of PEth 16:0/18:1 in blood is longer than that of ethyl glucuronide (EtG) or ethyl sulfate (EtS) in urine, measurement of blood PEth 16:0/18:1 level is routinely performed in clinical toxicology and forensic testing applications such as abstinence monitoring and workplace drug testing. PEth 16:0/18:1 has also been investigated as a biomarker to confirm prenatal alcohol exposure (PAE).
TRUSTED_SUSTAINABILITY

Specifications

CAS 67-56-1
Linear Formula C39H69D5O8P · NC3H10
Concentration 100 μg/mL in methanol (as free phosphate)
Formula Weight 767.12
Packaging Ampule
Quantity 1 mL

CERILLIANT is a registered trademark of Cerilliant Corporation

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.