Learn More
CERILLIANT™ (+/-)-Phenylpropanolamine Hydrochloride Solution, 1.0 mg/mL in Methanol, Sold by MilliporeSigma™ Supelco™
Shop All MilliporeSigma Supelco ProductsDescription
Specifications
Specifications
| CAS | 67-56-1, 154-41-6 |
| Melting Point | 194 to 196°C (lit.) |
| Linear Formula | C6H5CH(OH)CH(NH2)CH3 · HCl |
| MDL Number | MFCD00013019 |
| UN Number | UN 1230 - class 3 - PG 2 - Methanol, solution |
| Concentration | 1.0 mg/mL in methanol (as free base) |
| Synonym | DL-Norephedrine hydrochloride, Norephedrine hydrochloride, (+/-)-Phenylpropanolamine hydrochloride, erythro-2-Amino-1-phenyl-1-propanol hydrochloride, DL-erythro-2-Amino-1-phenyl-1-propanol hydrochloride, DL-Phenylpropanolamine hydrochloride |
| Formula Weight | 187.67 |
| Packaging | Ampule |
| Quantity | 1 mL |
| Show More |
CERILLIANT is a registered trademark of Cerilliant Corporation
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.