Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CERILLIANT™ Posaconazole-D4 Solution, 1.0 mg/mL in Methanol, Sold by MilliporeSigma™ Supelco™

Catalog No. 01001875 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-001-875 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-001-875 Supplier MilliporeSigma Supelco Supplier No. P1081ML
 Restricted Item
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Certified reference material

This stable-labeled internal standard is suitable for quantification of posaconazole levels in blood, serum, or plasma by HPLC or LC-MS/MS. The antifungal posaconazole is used to treat invasive and systemic fungal infections. As a result of significant drug interactions, therapeutic drug monitoring of posaconazole is recommended when administered with protease inhibitors (PIs) or non-nucleoside reverse transcriptase inhibitors (NNRTIs).
TRUSTED_SUSTAINABILITY

Specifications

CAS 67-56-1, 1133712-26-1
Linear Formula C37H38D4F2N8O4
MDL Number MFCD28899339
UN Number UN 1230 - class 3 - PG 2 - Methanol, solution
Concentration 1.0 mg/mL in methanol
Formula Weight 704.80
Packaging Ampule
Quantity 1 mL
Flash Point 9.7°C (49.5°F) - closed cup

CERILLIANT is a registered trademark of Cerilliant Corporation

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.