Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CERILLIANT™ Posaconazole Solution, 2.0 mg/mL in Methanol, Sold by MilliporeSigma™ Supelco™

Catalog No. 01001872 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-001-872 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-001-872 Supplier MilliporeSigma Supelco Supplier No. P1031ML
 Restricted Item
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Certified reference material

A certified Snap-N-Spike™ Solution suitable as starting material for calibrators, controls, or linearity standards in posaconazol therapeutic drug monitoring or clinical and diagnostic testing applications in patient blood, serum, or plasma samples by LC-MS/MS or HPLC. Sold under trade names Noxafil™ and Posanol, posaconazole is a triazole antifungal drug prescribed for treatment of invasive fungal infections in severely immunocompromised patients.
TRUSTED_SUSTAINABILITY

Specifications

CAS 67-56-1, 171228-49-2
Linear Formula C37H42F2N8O4
MDL Number MFCD00941162
UN Number UN 1230 - class 3 - PG 2 - Methanol, solution
Concentration 2.0 mg/mL in methanol
Formula Weight 700.78
Packaging Ampule
Quantity 1 mL
Flash Point 9.7°C (49.5°F)

CERILLIANT is a registered trademark of Cerilliant Corporation

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.