Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CERILLIANT™ Prednisolone Solution, 100 μg/mL in Acetonitrile, Sold by MilliporeSigma™ Supelco™

Catalog No. 01001885 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-001-885 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-001-885 Supplier MilliporeSigma Supelco Supplier No. P1211ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Certified reference material

Prednisolone is a synthetic glucocorticoid and derivative of cortisol which is used to treat a variety of inflammatory and auto-immune conditions. It is the active metabolite of the drug prednisone and is used especially in patients with liver failure, as these individuals are unable to metabolise prednisone into prednisolone. This Certified Spiking Solution™ is suitable for use as starting materials in the preparation of calibrators and controls for prednisolone testing methods by GC/MS or LC-MS/MS.
TRUSTED_SUSTAINABILITY

Specifications

CAS 75-05-8, 50-24-8
Linear Formula C21H28O5
MDL Number MFCD00003649
UN Number UN 1648 3 / PGII
Concentration 100 μg/mL in acetonitrile
Formula Weight 360.44
Packaging Ampule
Quantity 1 mL
Flash Point 2°C (35.6°F) - closed cup

CERILLIANT is a registered trademark of Cerilliant Corporation

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.