Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CERILLIANT™ Prednisone Solution, 100 μg/mL in Acetonitrile, Sold by MilliporeSigma™ Supelco™

Catalog No. 01001886 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-001-886 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-001-886 Supplier MilliporeSigma Supelco Supplier No. P1221ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Certified reference material

Calibration standard and quality control for calibration or identification of prednisone in biological samples for mass spectrometry-based clinical testing applications including therapeutic drug monitoring, endocrinology, cancer treatment, and confirming the cause of secondary adrenal insufficiency. Prednisone is a synthetic corticosteroid drug used to treat a variety of inflammatory diseases and conditions.
TRUSTED_SUSTAINABILITY

Specifications

CAS 75-05-8, 53-03-2
Melting Point -48°C
Linear Formula C21H26O5
MDL Number MFCD00003608
UN Number UN 1648 3 / PGII
Concentration 100 μg/mL in acetonitrile
Density 0.78 g/cm3 at 20°C
Formula Weight 358.43
Packaging Ampule
Quantity 1 mL
Flash Point 2°C (35.6°F) - closed cup
Show More Show Less

CERILLIANT is a registered trademark of Cerilliant Corporation

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.