Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CERILLIANT™ Promethazine-D3 Hydrochloride Solution, 100 μg/mL (1% 1 M HCl in Methanol), Sold by MilliporeSigma™ Supelco™

Catalog No. 01001878 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-001-878 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-001-878 Supplier MilliporeSigma Supelco Supplier No. P1121ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Certified reference material

A stable-labeled internal standard for promethazine testing or isotope dilution methods by GC/MS or LC/MS for clinical toxicology, urine drug testing, or forensic analysis. Promethazine is a phenothiazine analog prescribed for treatment of motion sickness, nausea, vomiting and allergies. Sold under the trade names Phenergan™ and Phenadoz™, promethazine has antihistamine, sedative, and mild antipsychotic effects. Illicit drug users have been reported to combine promethazine with other drugs such as opioids and benzodiazepines to enhance feelings of sedation.
TRUSTED_SUSTAINABILITY

Specifications

CAS 67-56-1, 1435933-86-0
Linear Formula C17H17D3N2S · HCl
UN Number UN 1230 - class 3 - PG 2 - Methanol, solution
Concentration 100 μg/mL (1% 1 M HCl in methanol (as free base))
Formula Weight 323.90
Packaging Ampule
Quantity 1 mL

CERILLIANT is a registered trademark of Cerilliant Corporation

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.