Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CERILLIANT™ Promethazine Hydrochloride Solution, 1.0 mg/mL (1% 1 M HCl in Methanol (as free base)), Sold by MilliporeSigma™ Supelco™

Catalog No. 01001877 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-001-877 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-001-877 Supplier MilliporeSigma Supelco Supplier No. P1111ML
 Restricted Item
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Certified reference material

Promethazine is a phenothiazine analog prescribed for treatment of nausea, motion sickness, vomiting, and allergies. Sold under trade names such as Phenergan™ and Phenadoz™, promethazine has antihistamine, sedative, and mild antipsychotic effects. This Certified Spiking Solution™ is suitable for GC/MS or LC/MS methods in promethazine testing applications including clinical toxicology, forensic analysis, and urine drug testing. Among drug abusers, promethazine has reportedly been misused in conjunction with opioids, benzodiazepines, and sedatives.
TRUSTED_SUSTAINABILITY

Specifications

CAS 67-56-1, 58-33-3
Linear Formula C17H20N2S · HCl
UN Number UN 1230 - class 3 - PG 2 - Methanol, solution
Concentration 1.0 mg/mL (1% 1 M HCl in methanol (as free base))
Formula Weight 320.88
Packaging Ampule
Quantity 1 mL

CERILLIANT is a registered trademark of Cerilliant Corporation

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.