Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CERILLIANT™ Propofol beta-D-glucuronide sodium salt, 100 μg/mL in Methanol (as free base), Sold by MilliporeSigma™ Supelco™

Catalog No. 01001857 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-001-857 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-001-857 Supplier MilliporeSigma Supelco Supplier No. P0821ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Certified reference material

Propofol glucuronide is the primary urinary metabolite of propofol, marketed as the anesthetic Diprivan™ for sedation and the induction and maintenance of general anesthesia. According to DEA's Drugs and Chemicals of Concern list, propofol abuse typically involves recreational use by anesthetists, practitioners, and other health care staff.
TRUSTED_SUSTAINABILITY

Specifications

CAS 67-56-1
Linear Formula C18H25NaO7
UN Number UN 1230 - class 3 - PG 2 - Methanol, solution
Concentration 100 μg/mL in methanol (as free base)
Synonym Propofol beta-D-glucuronide sodium salt solution
Formula Weight 376.38
Packaging Ampule
Quantity 1 mL
Flash Point 9.7°C (49.5°F) - closed cup

CERILLIANT is a registered trademark of Cerilliant Corporation

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.