Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CERILLIANT™ (+/-)-Propoxyphene-D5 Solution, 1.0 mg/mL in Acetonitrile, Sold by MilliporeSigma™ Supelco™

Catalog No. 01001905 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-001-905 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-001-905 Supplier MilliporeSigma Supelco Supplier No. P9041ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Certified reference material

Also known as dextropropoxyphene, propoxyphene is an opioid analgesic drug intended for treatment of mild pain. In combination with either paracetamol or aspirin, dextropropoxyphene is sold under trade names including Darvocet-N™ and Darvon™. This Certified Spiking Solution™ is suitable for use as an internal standard in propoxyphene testing methods by GC/MS or LC/MS for applications in urine drug testing, pain prescription monitoring, forensic analysis, or clinical toxicology.
TRUSTED_SUSTAINABILITY

Specifications

CAS 75-05-8, 136765-49-6
Linear Formula C22D5H24NO2
UN Number UN 1648 3 / PGII
Concentration 1.0 mg/mL in acetonitrile
Formula Weight 344.50
Packaging Ampule
Quantity 1 mL
Flash Point 2°C (35.6°F) - closed cup

CERILLIANT is a registered trademark of Cerilliant Corporation

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.